Learn More
Macherey-Nagel™ Chromafil™ Xtra PTFE Syringe Filters
Filter nonpolar liquids and gases. Chromafil™ Xtra PTFE Syringe Filters with polytetrafluoroethylene hydrophobic membrane.
Brand:  Macherey-Nagel™ 729207
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 10787548
  • 129.00 EUR / Pack of 100
Estimated Shipment: 21-05-2024
Description
  • Hydrophobic membrane
  • For nonpolar liquids and gases
  • Very resistant towards all kinds of solvents as well as acids and bases
  • Flushing with alcohol, followed by water, makes the originally hydrophobic membrane more hydrophilic
Specifications
Specifications
PTFE
∼4.9 cm2
100 Pack
∼80 μL
0.20 μm
Polypropylene (PP)
Syringe Filter
25 mm