Learn More
Macherey-Nagel™ Chromafil™ Xtra PTFE Syringe Filters
Filter nonpolar liquids and gases. Chromafil™ Xtra PTFE Syringe Filters with polytetrafluoroethylene hydrophobic membrane.
Brand: Macherey-Nagel™ 729205
Additional Details : Weight : 0.10000kg
Description
- Hydrophobic membrane
- For nonpolar liquids and gases
- Very resistant towards all kinds of solvents as well as acids and bases
- Flushing with alcohol, followed by water, makes the originally hydrophobic membrane more hydrophilic
Specifications
PTFE | |
∼4.9 cm2 | |
100 Pack | |
∼80 μL |
0.45 μm | |
Polypropylene (PP) | |
Syringe Filter | |
25 mm |