Learn More
Heger™ Polystyrene Petri Dishes
Brand:  Heger™ 1035
Additional Details : Weight : 7.20000kg
Product Code. 11361125
  • 143.00 EUR / Pack of 480
In Stock
Specifications
Clear
Polystyrene
Petri Dish
15 mm
Single-use
Round
480 Pack
90 mm