Learn More
Masterflex™ Polypropylene Straight Male Pipe Adapter
Compression fittings are an excellent choice to connect rigid tubing systems and withstand higher pressures than barbed fittings
Brand:  Masterflex™ 30815-24
Additional Details : Weight : 0.46000kg
 View more versions of this product
Product Code. 11760109
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • Assemble the fitting to the tubing by sliding the tubing through a sealing ferrule and using the nut to compress down to form a secure seal
  • PVDF Composition
Specifications
Specifications
Straight Male Pipe Adapter
Not Autoclavable
1 3/16 in.
1/4 in.
Ethylene Oxide
11/16 in.
Compression x NPT(M) Connection
Polypropylene
5/16 in.
5/16 in.
1/4 in.