Learn More
Masterflex™ Polypropylene Straight Male Pipe Adapter
Compression fittings are an excellent choice to connect rigid tubing systems and withstand higher pressures than barbed fittings
Brand: Masterflex™ 30815-24
Additional Details : Weight : 0.46000kg
Description
- Assemble the fitting to the tubing by sliding the tubing through a sealing ferrule and using the nut to compress down to form a secure seal
- PVDF Composition
Specifications
Straight Male Pipe Adapter | |
Not Autoclavable | |
1 3/16 in. | |
1/4 in. | |
Ethylene Oxide | |
11/16 in. |
Compression x NPT(M) Connection | |
Polypropylene | |
5/16 in. | |
5/16 in. | |
1/4 in. | |
White |