Learn More
Ismatec™ Reducer Tubing Connectors, Plastic
Variously sized and shaped reducer tubing connectors. Ismatec™ Reducer Tubing Connectors, Plastic are available in different plastic compositions.
31.38 EUR - 62.40 EUR
Specifications
Product Type | Reducer Tube Connector |
---|---|
Packaging Quantity | 10/Pk. |
Shape | Straight |
Product Code | Brand | Tubing Size | Shape | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Tubing Size | Shape | Price | Quantity & Availability | ||||
10308432
|
Ismatec™
ISM572 |
3.0mm and 4.0mm I.D. | Straight |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
10318432
|
Ismatec™
ISM575 |
6.0mm and 8.0mm I.D. | Straight |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
10554223
|
Ismatec™
ISM574 |
4.0mm and 8.0mm I.D. | Straight |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
10406992
|
Ismatec™
ISM570A |
1.5mm and 3.0mm I.D. | Straight |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
15295619
|
Ismatec™
ISM569A |
1.5mm and 2.5mm I.D. | Straight |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
10740325
|
Ismatec™
ISM571A |
2.5mm and 3.0mm I.D. | Straight |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
- Reducer tubing connector, with varying inlet and outlet diameters.
- Designed for Ismatec™ pump tubing.
- Available in different sizes, shapes, and compositions.
Specifications
Reducer Tube Connector | |
10/Pk. | |
Straight |