missing translation for 'onlineSavingsMsg'
Learn More

Socorex™ Polypropylene Pipette Tips

Use Socorex™ Polypropylene Pipette Tips when a whole volume range since there is a whole volume range from which to choose. They are available in various colors, are constructed of polypropylene and are autoclavable for cost effectiveness.

Brand:  Socorex™ 328.0200R

Additional Details : Weight : 0.80000kg

 View more versions of this product

Product Code. 11763266

  • 92.50 EUR / Pack of 960


Estimated Shipment
25-02-2021
Add to basket

Description

Description

  • Supplied in loose in bags or racks with lids
  • Autoclavable at 121°C / 250°F
  • Compatible with other brands' tips
Specifications

Specifications

10 μL
PP (Polypropylene)
Rack
Racked, 960/pk
Yellow
Socorex™, Eppendorf™
Universal
Videos
SDS
Documents