Learn More
Brady™ Polypropylene Operating Tool
Polypropylene Operating Tool which opens and locks the Pro-Lock II™ Cable Lockout Device.
11.10 EUR
Specifications
Product Type | Operating Tool |
---|---|
Dimensions (L x W) | 26 x 152 mm |
Quantity | 1 Ea. |
Product Code | Brand | Color | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Price | Quantity & Availability | |||||
11771975
|
Brady™
225207 |
Green |
11.10 EUR
Each |
Estimated Shipment: 03-05-2024
Log in to see stock. |
|||||
11761975
|
Brady™
225206 |
Red |
11.10 EUR
Each |
Estimated Shipment: 03-05-2024
Log in to see stock. |
|||||
11781975
|
Brady™
225208 |
Yellow |
11.10 EUR
Each |
Estimated Shipment: 03-05-2024
Log in to see stock. |
|||||
Description
- Operating tool which opens and locks the Pro-Lock II™ Cable Lockout Device.
Specifications
Operating Tool | |
26 x 152 mm | |
1 Ea. |