Learn More
Brady™ Polypropylene Mandatory Overshoes Signs
Brand: Brady™ 222643
Additional Details : Weight : 0.01300kg
Specifications
Use Of Overshoes Sign | |
Polypropylene | |
315 mm | |
120°C |
PPE Safety Sign | |
1 Ea. | |
Mandatory Use Of Overshoes Sign |