Learn More
Macherey-Nagel™ Polygram Alox N/UV254 Polyester Sheets
Macherey-Nagel Polygram Alox N/UV254 Polyester Sheets, Dimensions: 4 x 8 cm, Pore Size: 60 A, Fluorescent Indicator: UV254, Thickness: 0.2 mm
Brand:  Macherey-Nagel™ 802023
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 10499171
  • 147.00 EUR / Pack of 25
Estimated Shipment: 21-05-2024
Description
Macherey-Nagel Polygram Alox N/UV254 Polyester Sheets, Dimensions: 4 x 8 cm, Pore Size: 60 A, Fluorescent Indicator: UV254, Thickness: 0.2 mm
Specifications
Specifications
60 Å
Polyester Sheet
0.2 mm
25 Pk.
UV254
20 x 20 cm