missing translation for 'onlineSavingsMsg'
Learn More

Polyethylene glycol di-p-toluenesulfonate, M.W. 8,000, Thermo Scientific™

Catalog Number p-7080208
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
35164-96-6
C56H98O26S2
1251.49
JNHMMIVNUYBGER-UHFFFAOYSA-N
PEG-di-p-tosylate
62-[(4-methylbenzenesulfonyl)oxy]-3,6,9,12,15,18,21,24,27,30,33,36,39,42,45,48,51,54,57,60-icosaoxadohexacontan-1-yl 4-methylbenzene-1-sulfonate
CC1=CC=C(C=C1)S(=O)(=O)OCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOS(=O)(=O)C1=CC=C(C)C=C1
This item is not returnable. View return policy
35164-96-6
C56H98O26S2
1251.49
JNHMMIVNUYBGER-UHFFFAOYSA-N
PEG-di-p-tosylate
62-[(4-methylbenzenesulfonyl)oxy]-3,6,9,12,15,18,21,24,27,30,33,36,39,42,45,48,51,54,57,60-icosaoxadohexacontan-1-yl 4-methylbenzene-1-sulfonate
CC1=CC=C(C=C1)S(=O)(=O)OCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOS(=O)(=O)C1=CC=C(C)C=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Color White
Physical Form Granular Solid
Quantity 1 g
Chemical Name or Material Polyethylene glycol di-p-toluenesulfonate, M.W. 8,000
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'