Learn More
Cytiva Polyetheretherketone Tubing Fitting For SRTC-3 M6 Female
Shop All Cytiva Products
Click to view available options
Diameter (Metric) Inner:
1.5 mm
Unit Size:
Pack of 2
Specifications
Specifications
| Type | Tubing Fitting |
| Diameter (Metric) Outer | 1.8 mm |
| Material | PEEK |
| Diameter (Metric) Inner | 1.5 mm |
| For Use With (Equipment) | For SRTC-3 M6 Female |