Learn More
Brady™ Polyester Labels for Thermal Transfer Printers
For barcoding and identification purposes
Brand: Brady™ 116367
205.20 EUR valid until 2024-04-30
Use promo code "21673" to get your promotional price.
Additional Details : Weight : 0.48990kg
Description
- Available with gloss or matte finish
- Available in different widths and roll lengths
Specifications
RoHS 2002/95/EC | |
Acrylic, Permanent | |
Matte | |
Polyester | |
BBP™11 Label Printer | |
1500/Pk. | |
2.54 x 2.54 cm | |
0.048 mm | |
2.62 in. | |
1 in. | |
25.40 mm | |
2.54 cm |
Thermal Transfer Printable Labels | |
White, Translucent | |
Label | |
Laboratory identification such as vials, centrifuge tubes, test tubes, straws, and slides | |
1 | |
Rectangle | |
-196 °C to 110 °C | |
28.58 mm | |
2.54 cm | |
2.54 cm | |
1 in. |