missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polyanetholesulfonic Acid Sodium Salt, Reagent Grade, MP Biomedicals™
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Specifications
Color | White |
Quantity | 5 g |
Formula Weight | (250)n |
Grade | Reagent |
Physical Form | Solid |
Chemical Name or Material | Polyanetholesulfonic Acid Sodium Salt |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'