Learn More
Lenz Laborglasinstrumente™ Pointed Bottom Hollow Stoppers, Hexagonal Plate
For the safe and secure closure of flasks, bottles, tubing and other laboratory apparatus. Lenz Laborglasinstrumente™ Pointed Bottom Hollow Stoppers, Hexagonal Plate are manufactured from durable DURAN™ borosilicate glass.
Brand:  Lenz Laborglasinstrumente™ 01400014
Additional Details : Weight : 0.08100kg
 View more versions of this product
Product Code. 10261731
  • 16.64 EUR / Pack of 10
Estimated Shipment: 10-05-2024
Description
  • Acc. to DIN 12252, form C
  • Available in a range of cone sizes
  • Corrosion-free and resistant to thermal shock
Specifications
Specifications
Hollow
10/Pk.
Thread Size: 14/23 Cone NS
Hollow stopper
Borosilicate Glass