Learn More
BrandTech™ VITLAB™ PMP Wide Mouth Erlenmeyer Flasks
VITLAB wide-mouth Erlenmeyer flasks have screw caps for sample integrity
Brand: BrandTech™ 56995
Additional Details : Weight : 0.10000kg
Description
- Neck also accommodates NS stoppers
Specifications
Polypropylene | |
500 mL | |
100 mL | |
Wide | |
GL 52; NS 45/40 |
Transparent | |
PMP | |
6 | |
Screw |