Learn More
Alfa Aesar™ Platinum, 5% on activated carbon paste, Type 18
Brand:  Alfa Aesar™ H36968.22
Additional Details : CAS Number : 7440-06-4 Weight : 0.10000kg
Quantity 100g
 View more versions of this product
Product Code. 15459085
  • 977.00 EUR / 100g
Estimated Shipment date
from Supplier 04-06-2019
Add to basket
Specifications
Chemical Identifiers
7440-06-4
195.08
BASFCYQUMIYNBI-UHFFFAOYSA-N
23939
platinum
Pt
MFCD00011179
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
CHEBI:33400
[Pt]
Specifications
Platinum
7440-06-4
MFCD00011179
BASFCYQUMIYNBI-UHFFFAOYSA-N
platinum
23939
Solid
Gray
5% on activated carbon paste, Type 18
Pt
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
[Pt]
195.08
CHEBI:33400
Odorless
100g