Learn More
Kartell™ Plastilab™ Untapered T-Shaped Tubing Connectors
Designed with serrations to hold hoses tightly and prevent slipping.
Brand: Kartell™ 0046200
This item is not returnable.
View return policy
Description
- Made from polypropylene
- Autoclavable
- Chemical- and temperature-resistant
Compliance
Meet Food and Agriculture regulations
Specifications
Meet with the Food and Drug regulations. 90/128/CEE 92/39/CEE e FDA 21 CFR 177.1520, CE and IVD | |
Autoclavable | |
Suitable for foodstuff | |
6.3mm bore |
Untapered T Tubing Connector | |
Polypropylene | |
100/Pk. | |
T-shape |