Learn More
BRAND™ Plastic Type 3 Graduated Pipettes
Polypropylene, high clarity.
Brand:  BRAND™ 27610
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11728168
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • Calibrated to deliver (TD, Ex)
  • Resistant to breakage
  • Exposure to temperatures up to 60 °C will not cause permanent exceeding of error limits
  • Cleaning temperature below 60 °C is recommended to preserve marks and inscriptions
  • Specifications
    Specifications
    8 mm
    0.1 mL
    0.02 mL
    300 mm
    Polypropylene
    12
    2 mL