Learn More
Fisherbrand™ Plain PTFE Stir Bars
Provides stable stirring action. Fisherbrand™ Plain PTFE Stir Bars provide greater turbulence than cylindrical stir bars.
Brand:  Fisherbrand™
UNSPSC : 41120000
Additional Details : Weight : 0.00400kg
 View more versions of this product
Product Code. 10008241
  • 2.20 EUR / Each
In Stock 49
Add to basket
Specifications
Description
  • Stable, general-purpose magnetic stir bar
  • Provides greater turbulence than cylindrical stir bars
  • Autoclavable
Description & Specifications
Specifications
Yes
PTFE
PTFE encapsulated Alnico V magnet core
Containers & Vessels / Stirrer Hot Plates
20mm
Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
White
PTFE Stirrer Bars Plain
6mm
10 Pack