missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PKC-IN-1, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Description
PKC-IN-1 is a potent, ATP-competitive and reversible inhibitor of conventional PKC enzymes with Kis of 5.3 and 10.4 nM for human PKCβ and PKCα, and IC50s of 2.3, 8.1, 7.6, 25.6, 57.5, 314, 808 nM for PKCα, PKCβI, PKCβII, PKCθ, PKCγ, PKC mu and PKCε, respectively.
Specifications
Specifications
| Chemical Name or Material | PKC-IN-1 |
| Physical Form | Solid |
| CAS | 1046787-18-1 |
| Color | Off-White |
| Quantity | 5 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C25H37FN8O2 |
| For Use With (Application) | COVID-19-immunoregulation |
| Solubility Information | DMSO : ≥ 25 mg/mL (49.94 mM) |
| SMILES | O=C(N1[C@@H](C)CN(CC2CCOCC2)[C@H](C)C1)N(C3)C(C)(C)C4=C3C(NC5=NC(C)=NC=C5F)=NN4 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction