Learn More
Gilson™ PIPETMAN™ Classic 4-Pipette Kit
Shop All Gilson Products
Click to view available options
Volume (Metric):
0.2 to 1000 μL
Unit Size:
1 set
Description
- Provides exceptional accuracy and precision
- Easily adjusted volume can be trusted to exceed ISO recommendations when used with PIPETMAN™ Diamond tips
- Maintains consistent high level of performance for years to come with annual servicing
- PVDF body is resistant to aggressive chemicals and its piston assembly inhibits degradation of internal components
- Each kit includes: P2, P20, P200 and P1000 Classic pipettes, three TIPACK™ racks of Diamond tips, four SINGLE™ pipette holders, four PIPETMAN Comfort Handles, the Gilson™ guide to pipetting, and two-minute inspection poster
Specifications
Specifications
| Compatible Tips | Manual air displacement |
| Product Line | Pipetman Classic |
| Type | Pipette Kit |
| Volume (Metric) | 0.2 to 1000 μL |
| For Use With (Application) | Aspiration, Dispensing |
| Quantity | 1 |
| Resistance | Chemical resistance |
| Warranty | 3 years |