missing translation for 'onlineSavingsMsg'
Learn More

PIPES, ≥99%, Molecular Biology Reagent Grade, MP Biomedicals™

Catalog Number p-4405757
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
5625-37-6
C8H18N2O6S2
302.36
MFCD00006159
IHPYMWDTONKSCO-UHFFFAOYSA-N
Piperazine-N, N'-bis[2-ethanesulfonic acid]
79723
CHEBI:44933
2-[4-(2-sulfoethyl)piperazin-1-yl]ethanesulfonic acid
C1CN(CCN1CCS(=O)(=O)O)CCS(=O)(=O)O
This item is not returnable. View return policy
5625-37-6
C8H18N2O6S2
302.36
MFCD00006159
IHPYMWDTONKSCO-UHFFFAOYSA-N
Piperazine-N, N'-bis[2-ethanesulfonic acid]
79723
CHEBI:44933
2-[4-(2-sulfoethyl)piperazin-1-yl]ethanesulfonic acid
C1CN(CCN1CCS(=O)(=O)O)CCS(=O)(=O)O
Color White
Physical Form Powder
Quantity 25 g
Assay Percent Range ≥99%
Formula Weight 302.4
Grade Molecular Biology Reagent
Chemical Name or Material PIPES
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'