Learn More
ROBU VitraPOR™ Pipeline Filter
Pipeline Filter with tube adapters for connection.
Brand:  ROBU VitraPOR™ 23304
Additional Details : Weight : 0.09120kg
 View more versions of this product
Product Code. 16803056
  • 90.70 EUR / Pack of 2
Estimated Shipment: 31-05-2024
Description
  • Highly resistant to most chemicals and temperatures up to 500°C
Specifications
Specifications
Borosilicate Glass (3.3)
Tube Adapters
4
30 mm
Non-sterile
2/Pk.
140 mm