missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pilocarpine hydrochloride, 99.3%, MP Biomedicals™
Click to view available options
Quantity:
1 g
5 g
10 g
Specifications
Assay Percent Range | 0.993 |
Quantity | 1 g |
Formula Weight | 244.7 |
Physical Form | Crystalline |
Chemical Name or Material | Pilocarpine hydrochloride |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'