missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human PIK3CA/PIK3R1 (p110 alpha/p85 alpha), His Tag Recombinant Protein
Human PIK3CA/PIK3R1 (p110 alpha/p85 alpha), His Tag Recombinant Protein
870.00 EUR - 7890.00 EUR
Specifications
Gene Alias | PIK3CA/PIK3R1 (p110 alpha/p85 alpha), PI3 Kinase alpha |
---|---|
KinaseFamily | PI3 Kinase Family |
Species | Human |
Recombinant | Recombinant |
Expression System | Baculovirus |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15478673
|
Invitrogen™
PR8581C |
100μg |
1826.00 EUR
100µg |
Estimated Shipment: 15-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15458753
|
Invitrogen™
PV4789 |
1mg |
7890.00 EUR
1mg |
Estimated Shipment: 15-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10104653
|
Invitrogen™
PV4788 |
20 μg |
870.00 EUR
Each |
Estimated Shipment: 15-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
PIK3CA (p110 alpha) plays an important role in regulating cell growth and apoptosis. PIK3CA is coexpressed and copurified with the regulatory subunit PI3KR1.
Specifications
PIK3CA/PIK3R1 (p110 alpha/p85 alpha), PI3 Kinase alpha | |
Human | |
Baculovirus |
PI3 Kinase Family | |
Recombinant |