missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phosphomolybdotungstic Acid Reagent, Chem Lab™
Click to view available options
Quantity:
100 mL
Specifications
Color | Yellow |
Boiling Point | 100°C |
Melting Point | 0°C |
Composition | Na2WO4/Na2MoO4/HCl/H3PO4/Li2SO4/H2O |
Solubility Information | Water Soluble |
Odor | Odorless |
pH | <3 |
Quantity | 100 mL |
Physical Form | Liquid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'