missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phosphoenolpyruvic acid, trisodium salt hydrate, MP Biomedicals™
Click to view available options
Quantity:
25 mg
100 mg
250 mg
500 mg
1 g
5 g
Specifications
Color | White |
Quantity | 25 mg |
Formula Weight | 252 |
Physical Form | Crystalline Powder |
Chemical Name or Material | Phosphoenolpyruvic acid, trisodium salt hydrate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'