missing translation for 'onlineSavingsMsg'
Learn More

Phosphoenolpyruvic acid, trisodium salt hydrate, MP Biomedicals™

Catalog Number 11417092
Click to view available options
Quantity:
25 mg
100 mg
250 mg
500 mg
1 g
5 g
This item is not returnable. View return policy
5541-93-5
C3H2Na3O6P
233.986
RJUFBDMHZGRGMA-UHFFFAOYSA-K
2-propenoic acid, 2-phosphonooxy-, trisodium salt, trisodium 2-phosphonatooxy prop-2-enoate, unii-78y3q07j1x, phosphoenolpyruvate trisodium salt, 2-phosphoenol pyruvate na3salt, 2-propenoic acid, 2-phosphonooxy-, sodium salt 1:3, trisodium phosphonatoenolpyruvate, trisodium 2-phosphonatoacrylate, trisodium phosphoenolpyruvate, c3h2o6p.3na
79673
trisodium;2-phosphonatooxyprop-2-enoate
C=C(C(=O)[O-])OP(=O)([O-])[O-].[Na+].[Na+].[Na+]
This item is not returnable. View return policy
5541-93-5
C3H2Na3O6P
233.986
RJUFBDMHZGRGMA-UHFFFAOYSA-K
2-propenoic acid, 2-phosphonooxy-, trisodium salt, trisodium 2-phosphonatooxy prop-2-enoate, unii-78y3q07j1x, phosphoenolpyruvate trisodium salt, 2-phosphoenol pyruvate na3salt, 2-propenoic acid, 2-phosphonooxy-, sodium salt 1:3, trisodium phosphonatoenolpyruvate, trisodium 2-phosphonatoacrylate, trisodium phosphoenolpyruvate, c3h2o6p.3na
79673
trisodium;2-phosphonatooxyprop-2-enoate
C=C(C(=O)[O-])OP(=O)([O-])[O-].[Na+].[Na+].[Na+]
Color White
Quantity 25 mg
Formula Weight 252
Physical Form Crystalline Powder
Chemical Name or Material Phosphoenolpyruvic acid, trisodium salt hydrate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'