Learn More
Medicago Phosphate Buffered Saline Tablets with Tween™ 20, pH 7.4
Supplied as exactly pre-weighed tablets with a final buffer volume of 500 or 1000ml. Medicago Phosphate Buffered Saline Tablets with Tween™ 20, pH 7.4 is dissolved in deionized water to yield a solution of sodium chloride, potassium chloride, Tween™ 20 and phosphate buffer.
Brand:  Medicago 09-8902-100
Additional Details : Weight : 0.51120kg
 View more versions of this product
Product Code. 11310439
  • 198.00 EUR / Each
In Stock
Description
PBS-T buffer contains the non-ionic detergent Tween™ 20 which acts through blocking. It has the ability to reduce non-specific binding and protein-protein interaction during the wash step in protein and immunoassay procedures such as ELISA and Western blotting. Decreasing the non-specific binding and staining makes ELISA results and blots easier to interpret.
  • Formulated from analytical grade reagents
  • PBS with non-ionic detergent
  • Specifically developed for immunoassays
  • Exactly pre-weighed tablets
  • Dissolve and go for greater convenience
Specifications
Specifications
100 x 500 mL Tablets
ELISA, Western Blot, Plate-based Assays, Protein-plate Coating
7.4 at 25°C
Tablets
NaCl (0.14M), KCl (0.0027M), PO4 (0.01M), Tween™ 20 (0.05%)