Learn More
Köttermann Pharmacy Fume Hood without Media
This Pharmacy Fume Hood without Media is designed for the handling of hazardous substances in pharmacies and is ideally suited for working with dusty preparations or volatile chemicals.
Brand:  Köttermann 451-00018
Additional Details : Weight : 109.80810kg
 View more versions of this product
Product Code. 17382179
  • 2859.62 EUR / Each
Estimated Shipment: 04-09-2024
Description
  • For working safely with dust-forming preparations and volatile chemicals
  • Long-lasting steel construction with one-piece polyacrylic sash
  • Ergonomic features to improve workflow
  • Low, compact design for small laboratories
Specifications
Specifications
Ventilation Fan
Schuko Plug
115 kg
640 x 900 x 1350 mm
230 V