Learn More
pH 7.413 DIN 19266 Buffer Solution at 25°C, Reagecon
Tested, certified, NIST traceable, and shelf-stable buffer solution.
Brand: Reagecon 107413
This item is not returnable.
View return policy
Description
- pH Buffer Reference Standard, pH Buffer Reference Material
- Tested and certified by an ISO17025 Accredited Test Method (INAB Ref: 264T) and NIST traceable.
- Guaranteed stability throughout its entire shelf life, even after opening the bottle.
Specifications
Accuracy ± 0.010 pH units | |
Liquid | |
HDPE Bottle GL45 |
7.413 ±0.010 | |
500 mL |