missing translation for 'onlineSavingsMsg'
Learn More

Pentafluorophenyl 4-(1,3-oxazol-5-yl)benzoate, 97%, Maybridge™

110.60 EUR

Chemical Identifiers

CAS 898288-91-0
Molecular Formula C16H6F5NO3
Molecular Weight (g/mol) 355.22
MDL Number MFCD09025822
InChI Key FWUPBFHGEDUEID-UHFFFAOYSA-N
Synonym pentafluorophenyl 4-1,3-oxazol-5-yl benzoate, 2,3,4,5,6-pentafluorophenyl 4-1,3-oxazol-5-yl benzoate, perfluorophenyl 4-oxazol-5-yl benzoate, 4-5-oxazolyl benzoic acid 2,3,4,5,6-pentafluorophenyl ester, benzoic acid,4-5-oxazolyl-, 2,3,4,5,6-pentafluorophenyl ester, 2,3,4,5,6-pentakis fluoranyl phenyl 4-1,3-oxazol-5-yl benzoate
PubChem CID 18525706
IUPAC Name (2,3,4,5,6-pentafluorophenyl) 4-(1,3-oxazol-5-yl)benzoate
SMILES C1=CC(=CC=C1C2=CN=CO2)C(=O)OC3=C(C(=C(C(=C3F)F)F)F)F
View More Specs

Products 1
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10001434 SDS View Documents Maybridge
CC14526DA
1g Amber Glass Bottle 110.60 EUR
1g View alternative products
Save Up to 
»
Estimated Shipment
29-07-2020
Add to basket
 
Specifications

Chemical Identifiers

898288-91-0
355.22
FWUPBFHGEDUEID-UHFFFAOYSA-N
18525706
C1=CC(=CC=C1C2=CN=CO2)C(=O)OC3=C(C(=C(C(=C3F)F)F)F)F
C16H6F5NO3
MFCD09025822
pentafluorophenyl 4-1,3-oxazol-5-yl benzoate, 2,3,4,5,6-pentafluorophenyl 4-1,3-oxazol-5-yl benzoate, perfluorophenyl 4-oxazol-5-yl benzoate, 4-5-oxazolyl benzoic acid 2,3,4,5,6-pentafluorophenyl ester, benzoic acid,4-5-oxazolyl-, 2,3,4,5,6-pentafluorophenyl ester, 2,3,4,5,6-pentakis fluoranyl phenyl 4-1,3-oxazol-5-yl benzoate
(2,3,4,5,6-pentafluorophenyl) 4-(1,3-oxazol-5-yl)benzoate

Specifications

Pentafluorophenyl 4-(1, 3-oxazol-5-yl)benzoate
1g
96.5
97%
MFCD09025822
FWUPBFHGEDUEID-UHFFFAOYSA-N
(2,3,4,5,6-pentafluorophenyl) 4-(1,3-oxazol-5-yl)benzoate
18525706
97%
Amber Glass Bottle
898288-91-0
100.0
C16H6F5NO3
pentafluorophenyl 4-1,3-oxazol-5-yl benzoate, 2,3,4,5,6-pentafluorophenyl 4-1,3-oxazol-5-yl benzoate, perfluorophenyl 4-oxazol-5-yl benzoate, 4-5-oxazolyl benzoic acid 2,3,4,5,6-pentafluorophenyl ester, benzoic acid,4-5-oxazolyl-, 2,3,4,5,6-pentafluorophenyl ester, 2,3,4,5,6-pentakis fluoranyl phenyl 4-1,3-oxazol-5-yl benzoate
C1=CC(=CC=C1C2=CN=CO2)C(=O)OC3=C(C(=C(C(=C3F)F)F)F)F
355.22
355.22