Learn More
BRAND™ PEHD Double Stirring Spatula
PE-HD coated steel core
Brand:  BRAND™ 136100
Additional Details : Weight : 0.80000kg
Product Code. 12000922
  • 7.10 EUR / Each
Estimated Shipment: 28-05-2024
Description
  • Two differently shaped spatula ends
  • Approximately 16 mm wide
  • Specifications
    Specifications
    1
    PEHD
    260 mm
    Differently shaped ends
    Double Ended Spatula