Learn More
Duran™ PBT Screw Caps
Manufactured from Polybutylene terephthalate (PBT) DURAN™ PBT Screw Caps have excellent temperature and chemical resistance
13.55 EUR - 36.30 EUR
Products 2
Product Code Brand Description Price Quantity & Availability  
Product Code Brand Description Price Quantity & Availability  
11767198 View accessory products View Documents Duran™
292270602
Thread Size: GL 18 13.55 EUR
Pack of 10 View alternative products
Save Up to 
»
Estimated Shipment
04-07-2019
Add to basket
10434301 View accessory products View Documents Duran™
292271007
Thread Size: GL 45 36.30 EUR
Pack of 10 View alternative products
Save Up to 
»
In Stock 2
Add to basket
Description
Description
  • Excellent temperature and chemical resistance
  • Autoclavable
  • Open topped for septa or connectors