missing translation for 'onlineSavingsMsg'
Learn More

Palladium hydroxide, Pd 5% on carbon powder, Type A403002-5, nominally 50% water, Thermo Scientific Chemicals

Catalog Number p-7059901
Click to view available options
Quantity:
2 g
10 g
This item is not returnable. View return policy
12135-22-7
H2O2Pd
140.43
MFCD00064599
NXJCBFBQEVOTOW-UHFFFAOYSA-L
palladium hydroxide, palladium ii hydroxide, pearlman's catalyst, pearlmans catalyst, dihydroxypalladium, palladiumhydroxide, pearlman catalyst, peariman's catalyst, pearl man's catalyst, pd oh 2 on carbon
9942167
palladium(2+);dihydroxide
[OH-].[OH-].[Pd++]
This item is not returnable. View return policy
12135-22-7
H2O2Pd
140.43
MFCD00064599
NXJCBFBQEVOTOW-UHFFFAOYSA-L
palladium hydroxide, palladium ii hydroxide, pearlman's catalyst, pearlmans catalyst, dihydroxypalladium, palladiumhydroxide, pearlman catalyst, peariman's catalyst, pearl man's catalyst, pd oh 2 on carbon
9942167
palladium(2+);dihydroxide
[OH-].[OH-].[Pd++]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Black
Physical Form Powder
Quantity 2 g
Odor Odorless
Chemical Name or Material Palladium hydroxide, Pd 5% on carbon powder, Type A403002-5, Nominally 50% water

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'