missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Palladium, 5% on carbon, Type 87L, dry, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
Description
It is a catalyst used in Stille reaction, hydrogenation reactions, Suzuki-Miyaura cross-coupling and Heck-Mizoroki reactions, deoxygenation, oxidation reactions and coupling reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is a catalyst used in Stille reaction, hydrogenation reactions, Suzuki-Miyaura cross-coupling and Heck-Mizoroki reactions, deoxygenation, oxidation reactions and coupling reactions.
Solubility
Insoluble in water and organic solvents.
Notes
Palladium on carbon should always be handled under an inert atmosphere (preferably argon), and reaction vessels should be flushed with inert gas before the catalyst is added.
It is a catalyst used in Stille reaction, hydrogenation reactions, Suzuki-Miyaura cross-coupling and Heck-Mizoroki reactions, deoxygenation, oxidation reactions and coupling reactions.
Solubility
Insoluble in water and organic solvents.
Notes
Palladium on carbon should always be handled under an inert atmosphere (preferably argon), and reaction vessels should be flushed with inert gas before the catalyst is added.
Specifications
Quantity | 50 g |
UN Number | UN1325 |
Merck Index | 14,6989 |
Solubility Information | Insoluble in water and organic solvents. |
Chemical Name or Material | Palladium, 5% on carbon, Type 87L, dry |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'