missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Palladium, 5% on activated carbon paste, Type 424

Brand:  Alfa Aesar™ H37429.22

Additional Details : CAS Number : 7440-05-3 Weight : 0.10000kg

Quantity 100g

 View more versions of this product

Product Code. 15459095

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +358 9 8027 6280 for assistance
View alternative products »


Specifications

Chemical Identifiers

7440-05-3
106.42
KDLHZDBZIXYQEI-UHFFFAOYSA-N
CHEBI:33363
Pd
MFCD03457879
23938

Specifications

Palladium
7440-05-3
MFCD03457879
[Pd]
106.42
CHEBI:33363
Odorless
100g
5% on activated carbon paste, Type 424
Pd
KDLHZDBZIXYQEI-UHFFFAOYSA-N
palladium
23938
Solid
Gray
Videos
SDS