Learn More
p-Fluorotoluene, 97%, Thermo Scientific Chemicals
Catalog Number p-135897
Click to view available options
Quantity:
10 g
100 g
250 g
Packaging:
Glass bottle
352-32-9
MFCD00000358
WRWPPGUCZBJXKX-UHFFFAOYSA-N
4-fluorotoluene, p-fluorotoluene, benzene, 1-fluoro-4-methyl, p-fluoromethylbenzene, toluene, p-fluoro, 1-methyl-4-fluorobenzene, p-fluoromethyl benzene, unii-nud6wz1b8f, p-tolyl fluoride, para-fluorotoluene
9603
1-fluoro-4-methylbenzene
CC1=CC=C(C=C1)F
352-32-9
MFCD00000358
WRWPPGUCZBJXKX-UHFFFAOYSA-N
4-fluorotoluene, p-fluorotoluene, benzene, 1-fluoro-4-methyl, p-fluoromethylbenzene, toluene, p-fluoro, 1-methyl-4-fluorobenzene, p-fluoromethyl benzene, unii-nud6wz1b8f, p-tolyl fluoride, para-fluorotoluene
9603
1-fluoro-4-methylbenzene
CC1=CC=C(C=C1)F
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point -56°C
Color Colorless to Yellow
Density 1.0000g/mL
Boiling Point 116°C
Flash Point 10°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C7H7F
Linear Formula CH3C6H4F
Refractive Index 1.4674 to 1.4694
Quantity 10 g
Beilstein 05, 290
Merck Index 15, 4212
Specific Gravity 1
Solubility Information Solubility in water: insoluble.,soluble in alcohols,ether
Molecular Weight (g/mol) 110.13
Formula Weight 110.13
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material p-Fluorotoluene, 97%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • p-Fluorotoluene
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H335-May cause respiratory irritation.
  • H225-Highly flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P312-Call a POISON CENTER/doctor/physician/if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.