Learn More
Thermo Scientific Chemicals Oxytetracycline, 'can Be Used as Secondarystandard'
Catalog Number p-4510321
Click to view available options
Quantity:
500 mg
2.5 g
Packaging:
Glass Bottle
79-57-2
C22H26N2Na2O11
540.43
MFCD00003700
OLRBUPVCLOPDOT-GETZKYKJSA-M
54675779
(4S,4aR,5S,5aR,6S,12aR)-4-(dimethylamino)-1,5,6,10,11,12a-hexahydroxy-6-methyl-3,12-dioxo-4,4a,5,5a-tetrahydrotetracene-2-carboxamide
O.O.[Na+].[Na+].CN(C)[C@H]1[C@@H]2[C@@H](O)[C@H]3[C-](C(=O)C4=C([O-])C=CC=C4[C@@]3(C)O)C(=O)[C@]2(O)C(=O)\C(=C(\N)O)C1=O
79-57-2
C22H26N2Na2O11
540.43
MFCD00003700
OLRBUPVCLOPDOT-GETZKYKJSA-M
54675779
(4S,4aR,5S,5aR,6S,12aR)-4-(dimethylamino)-1,5,6,10,11,12a-hexahydroxy-6-methyl-3,12-dioxo-4,4a,5,5a-tetrahydrotetracene-2-carboxamide
O.O.[Na+].[Na+].CN(C)[C@H]1[C@@H]2[C@@H](O)[C@H]3[C-](C(=O)C4=C([O-])C=CC=C4[C@@]3(C)O)C(=O)[C@]2(O)C(=O)\C(=C(\N)O)C1=O
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Type Oxytetracycline
Physical Form Crystalline Powder
Melting Point 183.0°C
Infrared Spectrum Authentic
Packaging Glass Bottle
Solubility Information Solubility in water: 0.2g/L. Other solubilities: soluble in acids and alkalies, 25 mg/mL methanol, slightly soluble in acetone and chloroform, practically insoluble in ether
Formula Weight 460.43
Specific Rotation − 223.00
Percent Purity ≥80 wt% (as is, calculated) (HPLC)
Color Beige to Yellow
Quantity 500 mg
Chemical Name or Material Oxytetracycline
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Oxytetracycline
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 2
Hazard Statement
  • H361d-Suspected of damaging the unborn child.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.