missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OR2A4/OR2A7 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA538209
Additional Details : Weight : 0.00900kg
Description
Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms.Specifications
OR2A4/OR2A7 | |
Polyclonal | |
Unconjugated | |
OR2A7 | |
OR2A10 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
401427, 79541 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
O95047, Q96R45 | |
OR2A4, OR2A7 | |
A synthetic peptide derived from the internal region of human OR2A4/2A7 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |