Learn More
Hamilton™ Optional Locking Nut for Panel Mounting
For panel mounting of HVX valves.
Brand: Hamilton™ 35121
Additional Details : Weight : 0.01000kg
Description
- HVX valves are physically the largest valves
- Increased fluid flow
Specifications
Locking Nut | |
Panel Mounting | |
HVX Valves |