Learn More
Macherey-Nagel™ Optima™ XLB GC Capillary Column
Shop All Macherey-Nagel Products
Click to view available options
Length (Metric):
30 m
60 m
Unit Size:
Each
Specifications
Specifications
| Diameter (Metric) | 0.4 mm |
| For Use With (Equipment) | Ideal for Ion Trap and Quadrupole MS Detectors |
| Type | GC Capillary Column |
| Diameter (Metric) Inner | 0.25 mm |
| Stationary Phase | Optima™ XLB |
| For Use With (Application) | Gas Chromatography (GC) |
| Temperature Range | 340°C to 360°C |
| Film Thickness | 0.25 μm |
| Length (Metric) | 30 m |
| Quantity | 1 Each |