Learn More
Kolmi Hopen™ Op-Air™ Type II Medical Masks with Earloops
Shop All Medicom Products
Click to view available options
Color:
Blue
Green
Unit Size:
Pack of 2000
Pack of 60
Description
- High filtration media (polypropylene)
- Two wide and comfortable elastic bands
- Very soft polypropylene edges
- Anatomical folding limits exposure to face
- Integrated anti-fog nose bar
- Fiberglass-free filter
- Produced in ISO 8 clean room (particle and microbiological control)
Specifications
Specifications
| Color | Green |
| Fastening Type | Earloops |
| Material | Polypropylene |
| Style | Pleated |
| Certifications/Compliance (Europe) | CE, (EU) 2017/745, EN 14683:2019, AC:2019 |
| Disposable | Single-use |
| For Use With (Application) | Medical |
| Packaging Quantity | 60 Pk. |