Description
- Filtering half mask, efficiency class FFP2 NR D developed to protect against solid and liquid aerosols and particles
- Type IIR certified, it protects the environment against droplets emitted by the wearer and it protects from potential body fluid splashes
- Knitted headloops ultrasonically welded for optimal comfort
- Fits perfectly to face to limit inward leakage thanks to its shape and soft materials
- Hypoallergenic and has been dermatologically tested
- Packaged in individual polybags
- Made in France
Specifications
Specifications
| Certifications/Compliance | EN 149: 2001 + A1: 2009, DM Class I, EN 14683: 2019 + AC: 2019 |
| Color | White |
| Size | Small |
| Style | Pouch |
| Strap Attachment Type | Headband |