missing translation for 'onlineSavingsMsg'
Learn More

Octane, Reagent Grade, 98%, Honeywell™

Catalog Number p-7120638
Click to view available options
Quantity:
100 mL
500 mL
1 L
Packaging:
Glass Bottle
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
111-65-9
C8H18
114.232
MFCD00009556
TVMXDCGIABBOFY-UHFFFAOYSA-N
n-octane, oktan, oktanen, ottani, oktanen dutch, heptane, methyl, oktan polish, ottani italian, octane, all isomers, unii-x1rv0b2fjv
356
CHEBI:17590
octane
CCCCCCCC
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
111-65-9
C8H18
114.232
MFCD00009556
TVMXDCGIABBOFY-UHFFFAOYSA-N
n-octane, oktan, oktanen, ottani, oktanen dutch, heptane, methyl, oktan polish, ottani italian, octane, all isomers, unii-x1rv0b2fjv
356
CHEBI:17590
octane
CCCCCCCC

Reagent Grade, 98%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY
Melting Point -57°C
Density 0.703g/mL at 25°C
Boiling Point 125°C to 127°C
Flash Point 13°C (55.4°F)
Infrared Spectrum Conforms to structure
Packaging Glass Bottle
Linear Formula CH3(CH2)6CH3
Refractive Index n20/D 1.398
Quantity 100 mL
UN Number UN1262
Beilstein 1696875
Vapor Density 3.9 (vs air)
Formula Weight 114.23g/mol
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Octane
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'