Learn More
BRAND™ Octagonal Magnetic Stirring Bar
Octagonal Magnetic Stirring Bar constructed of PTFE and equipped with a pivot ring.
Brand: BRAND™ 137615
65.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Angular surface creates high turbulence even at low speeds
- For vessels with slightly uneven bottoms
- The pivot ring reduces the contact surface and enables the stirrer to adopt the optimum position
Specifications
Pivot Ring | |
38 mm | |
Octagon |
8 mm | |
PTFE |