Learn More
Motic™ Objective, Plan, Strain-free, CCIS EC for BA310
Color-corrected infinity system, enhanced-contrast Plan objective. Motic™ Objective, Plan, Strain-free, CCIS EC for BA310 is designed for field curvature correction and strain-free use.
Brand:  Motic™ 1101001703961
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15455272
  • 273.00 EUR / Each
Estimated Shipment: 01-08-2024
to see stock.
Add to basket
Description
Description
  • Suitable for BA310 Polarizing Microscope.
  • Color-corrected infinity system.
  • Enhanced contrast.
  • Field curvature correction.
  • Strain-free.
Specifications
Specifications
CCIS, EC Plan, Strain-free
NA: 0.40, WD: 0.9mm
BA310 Polarizing Microscope
20X