Learn More
Motic™ Objective, Plan, Strain-free, CCIS EC for BA310
Color-corrected infinity system, enhanced-contrast Plan objective. Motic™ Objective, Plan, Strain-free, CCIS EC for BA310 is designed for field curvature correction and strain-free use.
Brand: Motic™ 1101001703951
Additional Details : Weight : 0.01000kg
Descrizione
- Suitable for BA310 Polarizing Microscope.
- Color-corrected infinity system.
- Enhanced contrast. Field curvature correction.
- Strain-free.
Specifica
CCIS, EC Plan, Strain-free | |
NA: 0.25, WD: 17.4mm | |
BA310 Polarizing Microscope | |
10X |