Learn More
Motic™ Objective, CCIS Plan Achromatic NCG EC PL for BA310E
Color-corrected infinity system, enhanced-contrast, non-cover glass and lead-free objective. Motic™ Objective, CCIS Plan Achromatic NCG EC PL for BA310E is designed for field curvature, spherical aberration, and chromatic aberration correction.
63.55 EUR - 167.00 EUR
Specifications
Type Objective
For Use With (Equipment) BA310E Microscopes
View More Specs
Products 4
Product Code Brand Description Magnification Power Price Quantity & Availability  
Product Code Brand Description Magnification Power Price Quantity & Availability  
15445242
View Documents Promotion Details
Motic™
1101001702991
NA: 0.25, WD: 4.45mm 10X This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15455242
View Documents Promotion Details
Motic™
1101001703001
NA: 0.45, WD: 1mm 20X This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15465242
View Documents Promotion Details
Motic™
1101001703011
NA: 0.65/S, WD: 0.55mm 40X This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15475242
View Documents Promotion Details
Motic™
1101001703021
NA: 1.25/S-Oil, WD: 0.24 100X This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description
Description
  • Suitable for BA310 Series Upright Compound Microscope.
  • Color-corrected infinity system.
  • Non-cover glass system.
  • Lead-free glass.
  • Enhanced contrast.
  • Field curvature correction.
  • Chromatic aberration correction for 2 colors.
  • Spherical correction for 1 color.
Specifications
Specifications
Objective
BA310E Microscopes