Learn More
DWK Life Sciences O-Ring, transparent, from silicone (VMQ), not suitable for desiccators
Seal is suitable for the tight sealing of flat flange vessels with annular groove.
53.40 EUR - 217.00 EUR
Specifications
Material | Silicone (VMQ) |
---|---|
Quantity | 5/Pk. |
For Use With (Application) | Tight closure of flange vessels |
Product Code | Brand | Diameter (Metric) Outside | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Outside | Price | Quantity & Availability | |||||
11890680
|
DWK Life Sciences
292256106 |
215 mm |
217.00 EUR
Pack of 5 |
Estimated Shipment: 16-05-2024
Log in to see stock. |
|||||
15282119
|
DWK Life Sciences
292254602 |
110 mm |
85.70 EUR
Pack of 5 |
Estimated Shipment: 16-05-2024
Log in to see stock. |
|||||
13497068
|
DWK Life Sciences
292255101 |
133 mm |
126.00 EUR
Pack of 5 |
Estimated Shipment: 16-05-2024
Log in to see stock. |
|||||
11880680
|
DWK Life Sciences
292255701 |
157 mm |
170.00 EUR
Pack of 5 |
Estimated Shipment: 16-05-2024
Log in to see stock. |
|||||
11870680
|
DWK Life Sciences
292253409 |
75 mm |
53.40 EUR
Pack of 5 |
Estimated Shipment: 16-05-2024
Log in to see stock. |
|||||
Description
Flat flange vessels are large-volume reaction vessels consisting of a lower part and a lid. These two parts are held together by a clamping ring. The transparent O-ring made of silicone serves as a seal for flat flange vessels with an annular groove. It prevents tilting of the ground surfaces between the lower part and the lid and convinces by its good elasticity. The chemical resistance is limited compared to FEP coated O-rings.
- Accessories for flat flange vessels from silicone (VMQ), so is highly elastic
- The chemical resistance of silicone is lower than FEP coated O-rings
Specifications
Silicone (VMQ) | |
5/Pk. | |
Tight closure of flange vessels |